Tetrachlorosilane, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Silicon tetrachloride, Silicon (IV) chloride
M-Clarity™ quality level = MQ200
289388-100ML 289388-800ML
SIAL289388-100MLEA 107 EUR
SIAL289388-100ML SIAL289388-800ML
Tetrachlorosilane, Sigma-Aldrich®
Tetrachlorosilane
Formula: Cl₄Si
Boiling Pt: 57.6 °C (1013 hPa)
Melting Pt: –70 °C
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011229
CAS Number: 10026-04-7
EINECS: 233-054-0
UN: 1818
ADR: 8,II
Merck Index: 12,08644