Tin(II) chloride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Stannous chloride, Tin dichloride
452335-1G
452335-5G
SIAL452335-1GEA
171
EUR
SIAL452335-1G
SIAL452335-5G
Tin(II) chloride, Sigma-Aldrich®
Tin (II) chloride
Formula:
SnCl₂ Boiling Pt: 623 °C (1013 hPa) Melting Pt: 246 °C Density: 3.95 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011241 CAS Number: 7772-99-8 EINECS: 231-868-0 UN: 3260 ADR: 8,III Merck Index: 13,08861 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...