Tin(II) oxide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Stannous oxide, Tin monoxide, Tin(ii)oxide

M-Clarity™ quality level = MQ100

244643-100G 244643-500G
SIAL244643-100GEA 78.6 EUR
SIAL244643-100G SIAL244643-500G
Tin(II) oxide, Sigma-Aldrich®
Tin (II) oxide
Formula: OSn
MW: 134.71 g/mol
Boiling Pt: 1425
Melting Pt: 1040
Density: 6.45 g/cm³ (20 °C)
MDL Number: MFCD00011243
CAS Number: 21651-19-4
EINECS: 244-499-5
Merck Index: 12,08944

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR