Tin(IV) oxide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide
M-Clarity™ quality level = MQ100
204714-25G 204714-5G
SIAL204714-25GEA 597 EUR
SIAL204714-25G SIAL204714-5G
Tin(IV) oxide, Sigma-Aldrich®
Tin (IV) oxide
Formula: O₂Sn
MW: 150.71 g/mol
Boiling Pt: ∼2500 °C (1013 hPa)
Melting Pt: 1624.85
Density: 6.95 g/cm³ (20 °C)
MDL Number: MFCD00011244
CAS Number: 18282-10-5
EINECS: 242-159-0
Merck Index: 14,08778