Lead(II) sulphide (≥82% Pb)
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Lead sulphide
A10268.36 A10268.0E
A10268.36EA 52.6 EUR
A10268.36 A10268.0E
Lead(II) sulphide (≥82% Pb)
Lead (II) sulphide
CAS No.: 1314-87-0 Molecular Formula: PbS Formula Weight: 239.25 Refractive Index: 3.921 Odour: Odourless Melting Point: 1114° UN No.: 3077 MDL No.: MFCD00016280
Formula: PbS
Boiling Pt: 1344 °C (1013 hPa)
Melting Pt: 1114 °C
Density: 7.5
Storage Temperature: Ambient
MDL Number: MFCD00016280
CAS Number: 1314-87-0
EINECS: 215-246-6
UN: 3077
ADR: 9,III
Merck Index: 14,5421