Lead(II) sulphate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Lead sulphate
M-Clarity™ quality level = MQ200
307734-100G 307734-500G
SIAL307734-100GEA 39.1 EUR
SIAL307734-100G SIAL307734-500G
Lead(II) sulphate, Sigma-Aldrich®
Lead (II) sulphate
Formula: O₄PbS
MW: 303.26 g/mol
Melting Pt: 1000
Density: 6.0631 g/cm³ (18 °C)
MDL Number: MFCD00011166
CAS Number: 7446-14-2
EINECS: 231-198-9
UN: 2291
ADR: 6.1 III
Merck Index: 12,05444