(3-Aminopropyl)trimethoxysilane, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-(Trimethoxysilyl)propylamine, 3-(Trimethoxysilyl)propan-1-amine, Trimethoxy(3-aminopropyl)silane, 3-Aminopropyltrimethoxysilane
M-Clarity™ quality level = MQ200
281778-100ML 281778-500ML 281778-5ML
SIAL281778-100MLEA 93.7 EUR
SIAL281778-100ML SIAL281778-500ML SIAL281778-5ML
(3-Aminopropyl)trimethoxysilane, Sigma-Aldrich®
(3-Aminopropyl)trimethoxysilane
Formula: H₂NCH₂CH₂CH₂Si(OCH₃)₃
MW: 179.29 g/mol
Boiling Pt: 194
Density: 1.0265 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008206
CAS Number: 13822-56-5
EINECS: 237-511-5