N-(3-(Trimethoxysilyl)propyl)ethylenediamine, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 3-(2-Aminoethylamino)propyltrimethoxysilane, N-(2-Aminoethyl)-3-aminopropyltrimethoxysilane, N-[3-(Trimethoxysilyl)propyl]ethylenediamine

M-Clarity™ quality level = MQ100

440302-100ML 440302-500ML
SIAL440302-100MLEA 59.5 EUR
SIAL440302-100ML SIAL440302-500ML
N-(3-(Trimethoxysilyl)propyl)ethylenediamine, Sigma-Aldrich®
N-(3-(Trimethoxysilyl)propyl)ethylenediamine
Formula: H₂NCH₂CH₂NHCH₂CH₂CH₂Si(OCH₃)₃
MW: 222.36 g/mol
Boiling Pt: 261…263 °C (1013 hPa)
Melting Pt: ∼ 0 °C
Density: 1.01 g/cm³ (20 °C)
Flash Pt: 136
Storage Temperature: Ambient
MDL Number: MFCD00008173
CAS Number: 1760-24-3
EINECS: 217-164-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR