Dodecyltrimethoxysilane 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Trimethoxy(dodecyl)silane

L16486.14 L16486.22
L16486.14EA 54 EUR
L16486.14 L16486.22
Dodecyltrimethoxysilane 95%
Dodecyltrimethoxysilane

Purity: 95% CAS No.: 3069-21-4 Molecular Formula: CH3(CH2)11Si(OCH3)3 Formula Weight: 290.52 Flash Point: 108°(226°F) Refractive Index: 1.4274 Melting Point: -40° Boiling Point: 125°/1.8mm Beilstein Reference: 2356323 MDL No.: MFCD00069148

Formula: C₁₅H₃₄O₃Si
Boiling Pt: 125 °C (1.8 mmHg)
Melting Pt: –40 °C
Density: 0.890
Flash Pt: 108 °C (226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00069148
CAS Number: 3069-21-4
EINECS: 221-332-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR