Hexadecyltrimethoxysilane, Sigma-Aldrich®

Supplier: Merck

52360-100ML 52360-500ML
SIAL52360-100MLEA 45.1 EUR
SIAL52360-100ML SIAL52360-500ML
Hexadecyltrimethoxysilane, Sigma-Aldrich®
Hexadecyltrimethoxysilane
Formula: C₁₉H₄₂O₃Si
MDL Number: MFCD00069149
CAS Number: 16415-12-6
EINECS: 240-464-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR