Methyltriethoxysilane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
175579-250G 175579-50G
SIAL175579-250GEA 72.6 EUR
SIAL175579-250G SIAL175579-50G
Methyltriethoxysilane, Sigma-Aldrich®
Methyltriethoxysilane
Formula: C₇H₁₈O₃Si
Boiling Pt: 143…145 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: 30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009066
CAS Number: 2031-67-6
EINECS: 217-983-9