Dimethyldichlorosilane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Dichloro(dimethyl)silane, Dichlorodimethylsilane
MQ100
80430-25KG-F 80430-500G-F 80430-5KG-F
SIAL80430-25KG-FEA 1470 EUR
SIAL80430-25KG-F SIAL80430-500G-F SIAL80430-5KG-F
Dimethyldichlorosilane, Sigma-Aldrich®
Dimethyldichlorosilane
Formula: C₂H₆Cl₂Si
MW: 129.06 g/mol
Boiling Pt: 70
Melting Pt: –76 °C
Density: 1.0745 g/cm³ (20 °C)
Flash Pt: –16 °C
MDL Number: MFCD00000491
CAS Number: 75-78-5
EINECS: 200-901-0
UN: 1162
ADR: 3 II
REACH: 01-2119437250-51