Chlorotrimethylsilane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: TMCS, TMSC, Trimethylchlorosilane
95541-25KG-F 95541-500G-F
SIAL95541-25KG-FEA 2040 EUR
SIAL95541-25KG-F SIAL95541-500G-F
Chlorotrimethylsilane, Sigma-Aldrich®
Chlorotrimethylsilane
Packaging: produced by Wacker Chemie AG
Formula: C₃H₉ClSi
Boiling Pt: 59 °C (1013 hPa)
Melting Pt: –58 °C
Density: 0.8576 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00000502
CAS Number: 75-77-4
EINECS: 200-900-5
UN: 1298
ADR: 3,II
REACH: 01-2119457596-25