Tributyltin hydride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Tri-n-butyltin hydride
M-Clarity™ quality level = MQ100
234788-10G 234788-500G 234788-50G
SIAL234788-10GEA 62.5 EUR
SIAL234788-10G SIAL234788-500G SIAL234788-50G
Tributyltin hydride, Sigma-Aldrich®
Tributyltin hydride
Formula: C₁₂H₂₈Sn
MW: 291.06 g/mol
Boiling Pt: 112…114 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 109
MDL Number: MFCD00009416
CAS Number: 688-73-3
EINECS: 211-704-4
UN: 2929
ADR: 6.1 II