Triethylsilane, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
467448-100ML 467448-25ML
SIAL467448-100MLEA 196 EUR
SIAL467448-100ML SIAL467448-25ML
Triethylsilane, Sigma-Aldrich®
Triethylsilane
Formula: C₆H₁₆Si
MW: 116.28 g/mol
Boiling Pt: 107
Melting Pt: –156.9 °C
Density: 0.73 g/cm³ (20 °C)
Flash Pt: –3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009018
CAS Number: 617-86-7
EINECS: 210-535-3
UN: 1993
ADR: 3 II