Trimethylsilylacetylene, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Ethynyltrimethylsilane
M-Clarity™ quality level = MQ200
218170-1G
218170-25G
218170-5G
218170-100G
SIAL218170-1GEA
27.9
EUR
SIAL218170-1G
SIAL218170-25G
SIAL218170-5G
SIAL218170-100G
Trimethylsilylacetylene, Sigma-Aldrich®
Trimethylsilylacetylene
Formula:
C₅H₁₀Si Boiling Pt: 53 °C (1013 hPa) Density: 0.7111 g/cm³ (20 °C) Flash Pt: –34 °C |
MDL Number:
MFCD00008569 CAS Number: 1066-54-2 EINECS: 213-919-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...