1,2-Diphenyl-1,1,2,2-tetramethyldisilane 95%
Supplier: Thermo Fisher Scientific
Synonyms: 1,1,2,2-Tetramethyl-1,2-diphenyldisilane, 1,1,2,2-Tetramethyl-1,2-diphenyldisilane, 1,2-Diphenyltetramethyldisilane
H28705.06 H28705.03
H28705.06EA 147 EUR
H28705.06 H28705.03
1,2-Diphenyl-1,1,2,2-tetramethyldisilane 95%
1,2-Diphenyl-1,1,2,2-tetramethyldisilane
Purity: 97% CAS No.: 1145-98-8 Molecular Formula: C16H22Si2 Formula Weight: 270.52 MDL No.: MFCD00054786
Formula: C₁₆H₂₂Si₂
Boiling Pt: 111 °C
Melting Pt: 34…37 °C
Density: 0.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054786
CAS Number: 1145-98-8