Hexamethyldisilane ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A13155.14
A13155.22
A13155.14EA
101
EUR
A13155.14
A13155.22
Hexamethyldisilane ≥98%
Hexamethyldisilane
Purity: 98+%
CAS No.: 1450-14-2
Molecular Formula: (CH3)3SiSi(CH3)3
Formula Weight: 146.38
Flash Point: -1°(30°F)
Refractive Index: 1.4220
Odour: Odourless
Melting Point: 12-13°
Boiling Point: 112-113°
Beilstein Reference: 1633463
UN No.: 1993
MDL No.: MFCD00008258
Formula:
C₆H₁₈Si₂ Boiling Pt: 112…113 °C Melting Pt: 12…13 °C Density: 0.721 Flash Pt: –1 °C (30 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008258 CAS Number: 1450-14-2 EINECS: 215-911-0 UN: 1993 ADR: 3,II |