Chlorotriphenylsilane, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
114162-25G 114162-5G
SIAL114162-25GEA 77.2 EUR
SIAL114162-25G SIAL114162-5G
Chlorotriphenylsilane, Sigma-Aldrich®
Chlorotriphenylsilane
Formula: C₁₈H₁₅ClSi
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 90…95 °C
Density: 1.0165 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000496
CAS Number: 76-86-8
EINECS: 200-989-0
UN: 3261
ADR: 8,II