Triphenylsilane, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
148504-25G 148504-5G
SIAL148504-25GEA 113 EUR
SIAL148504-25G SIAL148504-5G
Triphenylsilane, Sigma-Aldrich®
Triphenylsilane
Formula: C₁₈H₁₆Si
Boiling Pt: 152 °C (1013 hPa)
Melting Pt: 43…45 °C
Density: 1.152 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003003
CAS Number: 789-25-3
EINECS: 212-333-0