Bismuth(III) bromide, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

401072-100G 401072-25G
SIAL401072-100GEA 176 EUR
SIAL401072-100G SIAL401072-25G
Bismuth(III) bromide, Sigma-Aldrich®
Bismuth (III) bromide
Formula: BiBr₃
MW: 448.69 g/mol
Boiling Pt: 278 °C (11 mmHg)
Melting Pt: 218
Density: 5.72 g/cm³ (20 °C)
MDL Number: MFCD00014195
CAS Number: 7787-58-8
EINECS: 232-121-1
UN: 3260
ADR: 8 III
Merck Index: 12,01299

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR