Phosphorus(III) bromide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Phosphorus tribromide
M-Clarity™ quality level = MQ200
157783-100G 157783-500G
SIAL157783-100GEA 89.5 EUR
SIAL157783-100G SIAL157783-500G
Phosphorus(III) bromide, Sigma-Aldrich®
Phosphorus (III) bromide
Formula: PBr₃
Boiling Pt: 175 °C (1013 hPa)
Melting Pt: –40 °C
Density: 2.8903 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011436
CAS Number: 7789-60-8
EINECS: 232-178-2
UN: 1808
ADR: 8,II
Merck Index: 12,07514