Phosphorus(V) bromide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Phosphorus pentabromide
M-Clarity™ quality level = MQ100
226734-100G 226734-25G
SIAL226734-100GEA 261 EUR
SIAL226734-100G SIAL226734-25G
Phosphorus(V) bromide, Sigma-Aldrich®
Phosphorus (V) bromide
Formula: PBr₅
Melting Pt: ? 100 °C
Density: 3.61 g/cm³ (20 °C)
MDL Number: MFCD00011437
CAS Number: 7789-69-7
EINECS: 232-186-6
UN: 2691
ADR: 8,II
Merck Index: 12,07507