Bismuth(III) chloride, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

450723-25G 450723-5G
SIAL450723-25GEA 383 EUR
SIAL450723-25G SIAL450723-5G
Bismuth(III) chloride, Sigma-Aldrich®
Bismuth (III) chloride
Formula: BiCl₃
Boiling Pt: 447 °C (1013 hPa)
Melting Pt: 232 °C
Density: 4.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003461
CAS Number: 7787-60-2
EINECS: 232-123-2
UN: 3260
ADR: 8,III
Merck Index: 13,01261

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR