Antimony(III) fluoride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Antimony trifluoride
M-Clarity™ quality level = MQ200
381292-100G
381292-500G
SIAL381292-100GEA
140
EUR
SIAL381292-100G
SIAL381292-500G
Antimony(III) fluoride, Sigma-Aldrich®
Antimony (III) fluoride
Formula:
F₃Sb Boiling Pt: 376 °C (1013 hPa) Melting Pt: 292 °C Density: 4.38 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011218 CAS Number: 7783-56-4 EINECS: 232-009-2 UN: 2923 ADR: 8,II Merck Index: 13,00715 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...