Bismuth(III) iodide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Bismuth triiodide
M-Clarity™ quality level = MQ100
341010-100G 341010-25G
SIAL341010-100GEA 372 EUR
SIAL341010-100G SIAL341010-25G
Bismuth(III) iodide, Sigma-Aldrich®
Bismuth (III) iodide
Formula: BiI₃
MW: 589.69 g/mol
Melting Pt: 408
Density: 5.78 g/cm³ (20 °C)
MDL Number: MFCD00010894
CAS Number: 7787-64-6
EINECS: 232-127-4
UN: 3260
ADR: 8 III
Merck Index: 12,01308