Bismuth(III) nitrate pentahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Bismuth trinitrate pentahydrate
M-Clarity™ quality level = MQ100
248592-100G 248592-500G
SIAL248592-100GEA 55.9 EUR
SIAL248592-100G SIAL248592-500G
Bismuth(III) nitrate pentahydrate, Sigma-Aldrich®
Bismuth (III) nitrate pentahydrate
Formula: Bi(NO₃)₃·5H₂O
MW: 485.07 g/mol
Boiling Pt: 75…80 °C (1013 hPa)
Melting Pt: 30
Density: 2.736 g/cm³ (20 °C)
MDL Number: MFCD00149157
CAS Number: 10035-06-0
EINECS: 233-791-8
UN: 1477
ADR: 5.1 II