Bismuth(III) oxide 98+%, pure
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Bismuth Trioxide, Dibismuth trioxide
192620010 192620050
ACRO192620010EA 142 EUR
ACRO192620010 ACRO192620050
Bismuth(III) oxide 98+%, pure
Bismuth (III) oxide
Formula: BiO₃
MW: 465.96 g/mol
Boiling Pt: 1890 °C (760 mmHg)
Melting Pt: 825 °C
Density: 8.9 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273
Order Now
Specification Test Results
Appearance Yellow Powder
Assay 98.0 to 102.0 %