Bismuth(III) oxide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Bismuth Trioxide, Dibismuth trioxide
M-Clarity™ quality level = MQ100
202827-10G 202827-250G 202827-50G
SIAL202827-10GEA 80.9 EUR
SIAL202827-10G SIAL202827-250G SIAL202827-50G
Bismuth(III) oxide, Sigma-Aldrich®
Bismuth (III) oxide
Formula: BiO₃
MW: 465.96 g/mol
Boiling Pt: 1890
Melting Pt: 825
Density: 8.9 g/cm³ (20 °C)
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273