Bismuth(III) oxide, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Bismuth Trioxide, Dibismuth trioxide

M-Clarity™ quality level = MQ100

637017-100G 637017-250G 637017-25G
SIAL637017-100GEA 251 EUR
SIAL637017-100G SIAL637017-250G SIAL637017-25G
Bismuth(III) oxide, Sigma-Aldrich®
Bismuth (III) oxide
Formula: BiO₃
MW: 465.96 g/mol
Boiling Pt: 1890
Melting Pt: 825
Density: 8.9 g/cm³ (20 °C)
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR