Triethyl phosphate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
538728-100ML 538728-1L 538728-4L
SIAL538728-100MLEA 59.1 EUR
SIAL538728-100ML SIAL538728-1L SIAL538728-4L
Triethyl phosphate, Sigma-Aldrich®
Triethyl phosphate
Formula: C₆H₁₅O₄P
MW: 182.16 g/mol
Boiling Pt: 215
Melting Pt: –57 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 116
Storage Temperature: Ambient
MDL Number: MFCD00009077
CAS Number: 78-40-0
EINECS: 201-114-5
Merck Index: 12,09806