Triphenyl phosphite, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

T84654-3KG T84654-500G T84654-5G
SIALT84654-3KGEA 252 EUR
SIALT84654-3KG SIALT84654-500G SIALT84654-5G
Triphenyl phosphite, Sigma-Aldrich®
Triphenyl phosphite
Formula: C₁₈H₁₅O₃P
MW: 310.29 g/mol
Boiling Pt: 360
Melting Pt: 22…25 °C
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 188
Storage Temperature: Ambient
MDL Number: MFCD00003032
CAS Number: 101-02-0
EINECS: 202-908-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR