Trioctylphosphine oxide ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A11062.14
A11062.22
A11062.36
A11062.14EA
47.2
EUR
A11062.14
A11062.22
A11062.36
Trioctylphosphine oxide ≥98%
Trioctylphosphine oxide
Purity: 98%
CAS No.: 78-50-2
Molecular Formula: C24H51OP
Formula Weight: 386.65
Flash Point: 252°(486°F)
Odour: Odourless
Boiling Point: 212-213°/0.3mm
Beilstein Reference: 1796648
MDL No.: MFCD00002083
Formula:
C₂₄H₅₁OP Boiling Pt: 212…213 °C (0.3 mmHg) Melting Pt: 52…57 °C Density: 0.88 Flash Pt: 252 °C (486 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002083 CAS Number: 78-50-2 EINECS: 201-121-3 |