Trioctylphosphine oxide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: TOPO, Tri-n-octylphosphine Oxide, Trioctylphosphinoxide
M-Clarity™ quality level = MQ200
223301-100G 223301-25G 223301-5G
SIAL223301-100GEA 167 EUR
SIAL223301-100G SIAL223301-25G SIAL223301-5G
Trioctylphosphine oxide, Sigma-Aldrich®
Trioctylphosphine oxide
Formula: C₂₄H₅₁OP
Boiling Pt: 409 °C (1013 hPa)
Melting Pt: 50…52 °C
Density: 0.88 g/cm³ (20 °C)
MDL Number: MFCD00002083
CAS Number: 78-50-2
EINECS: 201-121-3