Trioctylphosphine oxide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: TOPO, Tri-n-octylphosphine Oxide, Trioctylphosphinoxide
M-Clarity™ quality level = MQ100
346187-100G 346187-500G
SIAL346187-100GEA 74.3 EUR
SIAL346187-100G SIAL346187-500G
Trioctylphosphine oxide, Sigma-Aldrich®
Trioctylphosphine oxide
Formula: C₂₄H₅₁OP
MW: 386.64 g/mol
Boiling Pt: 409
Melting Pt: 50…52 °C
Density: 0.88 g/cm³ (20 °C)
MDL Number: MFCD00002083
CAS Number: 78-50-2
EINECS: 201-121-3