Cetrimonium bromide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide
M-Clarity™ quality level = MQ300
H5882-100G H5882-1KG H5882-500G
SIALH5882-100GEA 61.2 EUR
SIALH5882-100G SIALH5882-1KG SIALH5882-500G
Cetrimonium bromide, Sigma-Aldrich®
Cetrimonium bromide
Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364.45 g/mol
Melting Pt: 237…243 °C
Flash Pt: 244 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011772
CAS Number: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9 III
Merck Index: 13,02034