Cetrimonium bromide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide
M-Clarity™ quality level = MQ200
H6269-100G H6269-1KG H6269-250G H6269-500G
SIALH6269-100GEA 122 EUR
SIALH6269-100G SIALH6269-1KG SIALH6269-250G SIALH6269-500G
Cetrimonium bromide, Sigma-Aldrich®
Cetrimonium bromide
Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364.45 g/mol
Melting Pt: 237…243 °C
Flash Pt: 244 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011772
CAS Number: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9 III
Merck Index: 13,02034