Tetrabutylammonium bromide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
426288-100G 426288-25G
SIAL426288-100GEA 84 EUR
SIAL426288-100G SIAL426288-25G
Tetrabutylammonium bromide, Sigma-Aldrich®
Tetrabutylammonium bromide
Formula: C₁₆H₃₆BrN
MW: 322.37 g/mol
Melting Pt: 100…103 °C
Density: 1.148 g/cm³ (20 °C)
MDL Number: MFCD00011633
CAS Number: 1643-19-2
EINECS: 216-699-2