Tetraheptylammonium bromide, Sigma-Aldrich®
Supplier: Merck
Warning
87301-10G 87301-50G
SIAL87301-10GEA 81.1 EUR
SIAL87301-10G SIAL87301-50G
Tetraheptylammonium bromide, Sigma-Aldrich®
Tetraheptylammonium bromide
Formula: C₂₈H₆₀BrN
MW: 490.7 g/mol
Melting Pt: 96
MDL Number: MFCD00011861
CAS Number: 4368-51-8
EINECS: 224-459-3