Chlormequat chloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Chlorocholine chloride, 2-Chloroethyltrimethylammonium chloride, (2-Chloroethyl)trimethylammonium chloride
M-Clarity™ quality level = MQ200
C4049-25G C4049-5G
SIALC4049-25GEA 92.7 EUR
SIALC4049-25G SIALC4049-5G
Chlormequat chloride, Sigma-Aldrich®
Chlormequat chloride
Formula: ClCH₂CH₂N(CH₃)₃Cl
MW: 158.07 g/mol
Melting Pt: 243
Storage Temperature: Ambient
MDL Number: MFCD00011869
CAS Number: 999-81-5
EINECS: 213-666-4
UN: 2811
ADR: 6.1 III
Merck Index: 13,02120