Tetrabutylammonium chloride xhydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Tetrabutyl ammonium chloride xhydrate, Tetra-n-butylammonium chloride hydrate, Tetrabutylammonium chloride hydrate, TBAC
M-Clarity™ quality level = MQ200
345857-100G 345857-25G 345857-5G
SIAL345857-100GEA 241 EUR
SIAL345857-100G SIAL345857-25G SIAL345857-5G
Tetrabutylammonium chloride xhydrate, Sigma-Aldrich®
Tetrabutylammonium chloride hydrate
Formula: C₁₆H₃₆ClN·xH₂O
MW: 277.92 g/mol (anhy)
MDL Number: MFCD00011635
CAS Number: 37451-68-6
EINECS: 214-195-7