Tetrabutylammonium fluoride trihydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
86843-250G 86843-50G
SIAL86843-250GEA 598 EUR
SIAL86843-250G SIAL86843-50G
Tetrabutylammonium fluoride trihydrate, Sigma-Aldrich®
Tetrabutylammonium fluoride trihydrate
Formula: C₁₆H₃₆FN·3H₂O
Melting Pt: 62…63 °C
MDL Number: MFCD00149981
CAS Number: 87749-50-6
EINECS: 207-057-2
UN: 2811
ADR: 6.1,II