Acetylthiocholine iodide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (2-Mercaptoethyl)trimethylammonium iodide acetate, 2-(Acetylthio)-N,N,N-trimethylethan-1-aminium iodide, S-Acetylthiocholine iodide
M-Clarity™ quality level = MQ100
01480-1G 01480-25G 01480-5G
SIAL01480-1GEA 40.9 EUR
SIAL01480-1G SIAL01480-25G SIAL01480-5G
Acetylthiocholine iodide, Sigma-Aldrich®
Acetylthiocholine iodide
Formula: H₃CC(S)O(CH₂)₂N(CH₃)₃I
MW: 289.18 g/mol
Melting Pt: 204
Storage Temperature: Refrigerator
MDL Number: MFCD00011819
CAS Number: 1866-15-5
EINECS: 217-474-1
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9024633662ddbbc8a8efbcd594cd79dbbcbb2d6&vid=c8a063af769022ab97d866f5f7547b476c5949a7&prid=web&clid=SAPDAM