Acetylthiocholine iodide, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
(2-Mercaptoethyl)trimethylammonium iodide acetate, 2-(Acetylthio)-N,N,N-trimethylethan-1-aminium iodide, S-Acetylthiocholine iodide
M-Clarity™ quality level = MQ100
A5751-1G
A5751-250MG
A5751-5G
SIALA5751-1GEA
53.3
EUR
SIALA5751-1G
SIALA5751-250MG
SIALA5751-5G
Acetylthiocholine iodide, Sigma-Aldrich®
Acetylthiocholine iodide
Formula:
H₃CC(S)O(CH₂)₂N(CH₃)₃I MW: 289.18 g/mol Melting Pt: 204 Storage Temperature: Refrigerator |
MDL Number:
MFCD00011819 CAS Number: 1866-15-5 EINECS: 217-474-1 UN: 2811 ADR: 6.1 III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...