Ammonium tetrafluoroborate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Ammonium fluoroborate, ammonium fluoborate ammonium borofluoride ammonium fluoroborate [qr] ammonium tetrafluoroborate

M-Clarity™ quality level = MQ100

223727-100G 223727-500G
SIAL223727-100GEA 24.1 EUR
SIAL223727-100G SIAL223727-500G
Ammonium tetrafluoroborate, Sigma-Aldrich®
Ammonium tetrafluoroborate
Formula: H₄BF₄N
Melting Pt: 510 °C
Density: 1.87 g/cm³ (20 °C)
MDL Number: MFCD06245785
CAS Number: 13826-83-0
EINECS: 237-531-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR