Ammonium hydrogen carbonate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Ammonium bicarbonate
M-Clarity™ quality level = MQ200
09830-1KG 09830-500G
SIAL09830-1KGEA 110 EUR
SIAL09830-1KG SIAL09830-500G
Ammonium hydrogen carbonate, Sigma-Aldrich®
Ammonium hydrogen carbonate
Formula: NH₄HCO₃
Melting Pt: 35 °C
Density: 1.57 g/cm³ (20 °C)
MDL Number: MFCD00012138
CAS Number: 1066-33-7
EINECS: 213-911-5
Merck Index: 13,00497