Ammonium nitrate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
A3795-2.5KG
A3795-500G
SIALA3795-2.5KGEA
389
EUR
SIALA3795-2.5KG
SIALA3795-500G
Ammonium nitrate, Sigma-Aldrich®
Ammonium nitrate
Formula:
NH₄NO₃ Boiling Pt: 230 °C (1013 hPa) Melting Pt: 169 °C Density: 1.73 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011425 CAS Number: 6484-52-2 EINECS: 229-347-8 UN: 1942 ADR: 5.1,III Merck Index: 13,00538 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...