Diammonium hydrogen orthophosphate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
M-Clarity™ quality level = MQ200
09839-1KG 09839-250G 09839-50G
SIAL09839-1KGEA 279 EUR
SIAL09839-1KG SIAL09839-250G SIAL09839-50G
Diammonium hydrogen orthophosphate, Sigma-Aldrich®
di-Ammonium hydrogen orthophosphate
Formula: (NH₄)₂HPO₄
MW: 132.06 g/mol
Melting Pt: 185
Density: 1.619 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 13,00546