Ammonium dihydrogen phosphate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

M-Clarity™ quality level = MQ100

204005-100G 204005-25G 204005-500G
SIAL204005-100GEA 241 EUR
SIAL204005-100G SIAL204005-25G SIAL204005-500G
Ammonium dihydrogen phosphate, Sigma-Aldrich®
Ammonium dihydrogen phosphate
Formula: NH₄H₂PO₄
Melting Pt: 190 °C
Density: 1.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5
Merck Index: 13,00547

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR