Ammonium perrhenate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
316954-25G 316954-5G
SIAL316954-25GEA 531 EUR
SIAL316954-25G SIAL316954-5G
Ammonium perrhenate, Sigma-Aldrich®
Ammonium perrhenate
Formula: H₄NO₄Re
Density: 3.97 g/cm³ (20 °C)
MDL Number: MFCD00012377
CAS Number: 13598-65-7
EINECS: 237-075-6
UN: 1479
ADR: 5.1,II